CO1H324A7086 CombiPhos Catalysts Inc

5-Bromo-2-aminophenylboronic acid pinacol ester

  • Cas Number : 1558927-15-3
  • Molecular Weight : 297.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart