CO1H324A708B CombiPhos Catalysts Inc

2-Bromo-4-cyanophenylboronic acid pinacol ester

  • Cas Number : 2223055-37-4
  • Molecular Weight : 307.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart