CO1H324A708D CombiPhos Catalysts Inc

4-CHLORO-2-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-BENZONITRILE

  • Cas Number : 863868-19-3
  • Molecular Weight : 263.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart