CO1H324A7098 CombiPhos Catalysts Inc

5-(2-Methoxyphenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402227-46-6
  • Molecular Weight : 316.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart