CO1H324A709A CombiPhos Catalysts Inc

5-(2-Fluorophenyl)furan-2-boronic acid pinacol ester

  • Cas Number : 1402240-86-1
  • Molecular Weight : 288.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart