CO1H324A70B2 CombiPhos Catalysts Inc

methyl 3-(piperidin-4-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • Cas Number : 2223051-74-7
  • Molecular Weight : 345.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart