CO1H324A70BB CombiPhos Catalysts Inc

4-(Cyclopentyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 2223055-39-6
  • Molecular Weight : 278.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart