CO1H324A70C0 CombiPhos Catalysts Inc

3-(iso-Butyl)-5-(tert-butyl)phenylboronic acid pinacol ester

  • Cas Number : 2223043-95-4
  • Molecular Weight : 316.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart