CO1H324A70C3 CombiPhos Catalysts Inc

3-Cyclopropyl-5-(imidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223046-92-0
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart