CO1H324A70C4 CombiPhos Catalysts Inc

4-(4-Tolyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1040281-87-5
  • Molecular Weight : 300.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart