CO1H324A70C7 CombiPhos Catalysts Inc

4-(4-Fluorophenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402174-26-8
  • Molecular Weight : 304.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart