CO1H324A70C9 CombiPhos Catalysts Inc

2-Cyclopropyl-5-(1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223055-40-9
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart