CO1H324A70CE CombiPhos Catalysts Inc

3-(Pyrrolidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223006-37-7
  • Molecular Weight : 273.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart