CO1H324A70CF CombiPhos Catalysts Inc

4-(Cyclopentyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2096330-75-3
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart