CO1H324A70D6 CombiPhos Catalysts Inc

4-(4-Tolyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2096339-63-6
  • Molecular Weight : 284.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart