CO1H324A70DC CombiPhos Catalysts Inc

2-Amino-3-(N-Boc-pyrrolidin-3-yl)phenylboronic acid pinacol ester

  • Cas Number : 2222996-70-3
  • Molecular Weight : 388.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart