CO1H324A70DD CombiPhos Catalysts Inc

2-Cyclopropyl-4-(1H-pyrazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223048-98-2
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart