CO1H324A70E2 CombiPhos Catalysts Inc

5-Bromo-2-(1H-imidazol-1-yl)phenylboronic acid pinacol ester

  • Cas Number : 2223051-89-4
  • Molecular Weight : 349.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart