CO1H324A7988 CombiPhos Catalysts Inc

2-Bromo-3-(cyclopentyl)thiophene

  • Cas Number : 1034138-14-1
  • Molecular Weight : 231.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart