CO1H324A84DE CombiPhos Catalysts Inc

4-Fluoro-2-(3-fluorophenyl)pyridine

  • Cas Number : 1214392-13-8
  • Molecular Weight : 191.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart