CO1H324A8647 CombiPhos Catalysts Inc

2-Chloro-4-(3-tolyl)-1,3,5-triazine

  • Cas Number : 1417519-21-1
  • Molecular Weight : 205.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart