CO1H324A9B5C CombiPhos Catalysts Inc

tert-butyl 3-(2-fluoropyridin-4-yl)azetidine-1-carboxylate

  • Cas Number : 2093427-33-7
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart