CO1H324A9B68 CombiPhos Catalysts Inc

2-fluoro-4-(pyrrolidin-3-yl)pyridine

  • Cas Number : 1260819-28-0
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart