CO1H324A9C0A CombiPhos Catalysts Inc

2-Hydroxy-5-(2-thienyl)pyrimidine

  • Cas Number : 1111113-73-5
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart