CO1H324A9C14 CombiPhos Catalysts Inc

2-Chloro-5-(3-furyl)pyrimidine

  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart