CO1H324A9CB6 CombiPhos Catalysts Inc

2-Bromo-5-(2-fluorolphenyl)thiazole

  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart