CO1H324A9D45 CombiPhos Catalysts Inc

2-Bromo-5-(2-chlorophenyl)thiazole

  • Molecular Weight : 274.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart