CO1H324A9DEA CombiPhos Catalysts Inc

4-Bromo-1-(2-methyl-4-fluorophenyl)pyrazole

  • Molecular Weight : 255.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart