CO1H324A9E26 CombiPhos Catalysts Inc

2,4-Dichloro-6-(iso-propyl)-1,3,5-triazine

  • Cas Number : 30894-74-7
  • Molecular Weight : 192.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart