CO1H324A9E8A CombiPhos Catalysts Inc

1-(4-Chloro-2-pyridyl)-4-bromoimidazole

  • Cas Number : 1353855-82-9
  • Molecular Weight : 258.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart