CO1H324A9EA0 CombiPhos Catalysts Inc

1-(5-Bromothiazol-2-yl)-4-aminoimidazole

  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart