CO1H324A9EE7 CombiPhos Catalysts Inc

2-Amino-4-chloro-6-(3-thienyl)-1,3,5-triazine

  • Molecular Weight : 212.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart