CO1H324A9EF9 CombiPhos Catalysts Inc

2-Amino-4-chloro-6-(4-methoxyphenyl)pyrimidine

  • Cas Number : 688782-45-8
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart