CO1H324A9F16 CombiPhos Catalysts Inc

3-Bromo-1-(4-methoxyphenyl)pyrazole

  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart