CO1H324A9F83 CombiPhos Catalysts Inc

2,4-Dichloro-6-(5-iso-propyl-2-thienyl)-1,3,5-triazine

  • Molecular Weight : 274.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart