CO1H324A9FC1 CombiPhos Catalysts Inc

4,5-Dibromo-2-(3-fluorophenyl)thiazole

  • Molecular Weight : 337.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart