CO1H324AACF5 CombiPhos Catalysts Inc

(2-hydroxy-6-isobutylpyridin-4-yl)boronic acid

  • Cas Number : 2256747-23-4
  • Molecular Weight : 195.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart