CO1H324AACFD CombiPhos Catalysts Inc

(2-hydroxy-6-(tetrahydro-2H-pyran-4-yl)pyridin-4-yl)boronic acid

  • Cas Number : 2256747-31-4
  • Molecular Weight : 223.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart