CO1H324AAD19 CombiPhos Catalysts Inc

(2-(diethylamino)-6-hydroxypyridin-4-yl)boronic acid

  • Cas Number : 2256747-58-5
  • Molecular Weight : 210.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart