CO1H324AAD24 CombiPhos Catalysts Inc

(2-amino-6-fluoropyridin-4-yl)boronic acid

  • Cas Number : 1214879-87-4
  • Molecular Weight : 155.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart