CO1H324AAD36 CombiPhos Catalysts Inc

(3-cyano-4-(1H-pyrazol-1-yl)phenyl)boronic acid

  • Cas Number : 2256747-83-6
  • Molecular Weight : 213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart