CO1H324AAD8B CombiPhos Catalysts Inc

2-(4-bromo-1H-pyrazol-1-yl)-4-fluoropyridine

  • Cas Number : 2256748-12-4
  • Molecular Weight : 242.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart