CO1H324AADA0 CombiPhos Catalysts Inc

5-fluoro-2-(1H-imidazol-2-yl)pyridine

  • Cas Number : 1256794-92-9
  • Molecular Weight : 163.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart