CO1H324AADAF CombiPhos Catalysts Inc

4-bromo-6-(1H-imidazol-2-yl)pyrimidine

  • Cas Number : 2256748-30-6
  • Molecular Weight : 225.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart