CO1H324AADC0 CombiPhos Catalysts Inc

methyl 5-bromo-2-(1H-imidazol-2-yl)benzoate

  • Cas Number : 2256748-37-3
  • Molecular Weight : 281.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart