CO1H324AADC7 CombiPhos Catalysts Inc

2-(4-bromo-3-fluorophenyl)-1H-imidazole

  • Cas Number : 1370601-04-9
  • Molecular Weight : 241.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart