CO1H324AADEC CombiPhos Catalysts Inc

methyl 5-bromo-2-(3-methylpiperidin-1-yl)benzoate

  • Cas Number : 2256748-60-2
  • Molecular Weight : 312.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart