CO1H324AADEF CombiPhos Catalysts Inc

methyl 5-bromo-2-(4-methylpiperazin-1-yl)benzoate

  • Cas Number : 1131587-80-8
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart