CO1H324AADF9 CombiPhos Catalysts Inc

5-bromo-2-(2-methyl-1H-imidazol-1-yl)benzonitrile

  • Cas Number : 1770229-96-3
  • Molecular Weight : 262.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart