CO1H324AAE7A CombiPhos Catalysts Inc

4-bromo-1-cyclopropyl-1H-imidazol-2-amine

  • Cas Number : 1872068-72-8
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart